Learn More
Cytek Biosciences Inc cFluor BYG750 aHu CD183 Anti
Supplier: Cytek Biosciences Inc R720777
Product Name: cFluor BYG750 Anti-Human CD183 (CXCR3) (CEW33D)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG750 Anti-Human CD183 (CXCR3) (CEW33D)
Cytek Catalog Number: R7-20777
Marker: CD183 (CXCR3)
Target Species: Human
Clonality: Monoclonal
Clone: CEW33D
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor BYG750
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.