Learn More
Cytek Biosciences Inc cFluor R685 aHu CD56 Anti
Supplier: Cytek Biosciences Inc R720343
Product Name: cFluor R685 Anti-Human CD56 (TULY56)
Product Class: Antibody
Type: Primary
Product Name: cFluor R685 Anti-Human CD56 (TULY56)
Cytek Catalog Number: R7-20343
Marker: CD56
Target Species: Human
Clonality: Monoclonal
Clone: TULY56
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R685
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.