Learn More
Cytek Biosciences Inc cFluor UV515 aHu CD4 Anti
Supplier: Cytek Biosciences Inc R720853
Product Name: cFluor UV515 Anti-Human CD4 (RPA-T4)
Product Class: Antibody
Type: Primary
Product Name: cFluor UV515 Anti-Human CD4 (RPA-T4)
Cytek Catalog Number: R7-20853
Marker: CD4
Target Species: Human
Reported Cross Reactivity: Chimpanzee
Clonality: Monoclonal
Clone: RPA-T4
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor UV515
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.