Learn More
Cytek Biosciences Inc cFluor UV570 aMu CD161 Anti
Supplier: Cytek Biosciences Inc R720889
Product Name: cFluor UV570 Anti-Mouse CD161 (NK1.1) (PK136)
Product Class: Antibody
Type: Primary
Product Name: cFluor UV570 Anti-Mouse CD161 (NK1.1) (PK136)
Cytek Catalog Number: R7-20889
Marker: CD161 (NK1.1)
Target Species: Mouse
Clonality: Monoclonal
Clone: PK136
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor UV570
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.