Learn More
Cytek Biosciences Inc cFluor V435 aHu CD28 Anti
Supplier: Cytek Biosciences Inc R721349
Product Name: cFluor V435 Anti-Human CD28 (CD28.2)
Product Class: Antibody
Type: Primary
Product Name: cFluor V435 Anti-Human CD28 (CD28.2)
Cytek Catalog Number: R7-21349
Marker: CD28
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque
Clonality: Monoclonal
Clone: CD28.2
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V435
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.