Learn More
Cytek Biosciences Inc cFluor V450 aHu CD16 Anti
Supplier: Cytek Biosciences Inc R720184
Product Name: cFluor V450 Anti-Human CD16 (3G8)
Product Class: Antibody
Type: Primary
Product Name: cFluor V450 Anti-Human CD16 (3G8)
Cytek Catalog Number: R7-20184
Marker: CD16
Target Species: Human
Reported Cross Reactivity: Baboon|Capuchin Monkey|Chimpanzee|Common Marmoset|Cynomolgus Monkey|Pigtail Monkey|Rhesus Macaque|Sooty Mangabey|Squirrel Monkey
Clonality: Monoclonal
Clone: 3G8
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V450
Size: 25 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.