Learn More
Cytek Biosciences Inc cFluor V780 aMu CD4 Anti
Supplier: Cytek Biosciences Inc R720711
Product Name: cFluor V780 Anti-Mouse CD4 (RM4-5)
Product Class: Antibody
Type: Primary
Product Name: cFluor V780 Anti-Mouse CD4 (RM4-5)
Cytek Catalog Number: R7-20711
Marker: CD4
Target Species: Mouse
Clonality: Monoclonal
Clone: RM4-5
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor V780
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.