Learn More
Enzo Life Sciences CFTR monoclonal antibody (CF3) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX804214R100
Encompass_Preferred
Clone: CF3. Isotype: IgM. Immunogen: Synthetic peptide corresponding to aa 103-117(G103RIIASYDPDNKEER117) in the first extracellular loop of human and rabbit CFTR (cystic fibrosis transmembrane conductance regulator). This sequence is highly conserved in mouse, sheep, bovine and Xenopus laevis CFTR. Species reactivity: Human, Mouse. Applications: ICC, IP, WB. Formulation: Liquid. Diluted ascites containing 0.05% sodium azide.
Catalog No. 50-200-9831
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.