Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CFTR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1935
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1935 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1935 Supplier Invitrogen™ Supplier No. PA1935
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-935 detects cystic fibrosis transmembrane conductance factor (CFTR) from cells overexpressing the human protein. PA1-935 has been successfully used in immunocytochemistry procedures. Immunocytochemical staining of HEK293 cells overexpressing human CFTR with this antibody results in staining primarily of the plasma membrane. PA1-935 immunizing peptide corresponds to amino acid residues 103-117 from human CFTR protein. This sequence is completely conserved between human, rabbit, and monkey and there is a one amino acid substitution in rat, bovine, and sheep.

Cystic Fibrosis (CF) is a common lethal genetic disease caused by mutations of the gene coding for the cystic fibrosis transmembrane conductance factor, a cAMP regulated chloride channel. Approximately 70% of all CF cases share the deletion of a phenylalanine at position 508 (delta F508) which results in abnormal chloride transport. Since the CF mutation is lethal, most often by lung and liver disease, it raises the question of why this genetic disease remains as common as it is. One possible explanation is that Salmonella typhi has been shown to use CFTR to enter intestinal epithelial cells and that delta F508 heterozygote and homozygote mice showed 86% and 100% reductions in S. typhi intestinal submucosal uptake.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFTR
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene CFTR
Gene Accession No. P13569
Gene Alias ABC35; Abcc7; ATP Binding Cassette Superfamily C Member 7 (ABCC7); ATP-binding cassette sub-family C member 7; ATP-binding cassette transporter sub-family C member 7; ATP-binding cassette, subfamily c, member 7; AW495489; cAMP-dependent chloride channel; CF; CFTR; CFTR/MRP; Channel conductance-controlling ATPase; cystic fibrosis transmembrane conductance regulator; cystic fibrosis transmembrane conductance regulator (ATP-binding cassette sub-family C, member 7); cystic fibrosis transmembrane conductance regulator homolog; cystic fibrosis transmembrane conductance regulator homolog; ATP-binding cassette, subfamily c, member 7; dJ760C5.1; MRP7; RGD1561193; tcag7.78; TNR CFTR; TNR-CFTR
Gene Symbols CFTR
Host Species Rabbit
Immunogen Synthetic Peptide: G(103) R I I A S Y D P D N K E E R(117).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1080
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.