Learn More
Cole-Parmer™ CG-900 Cryo-Blade™ Cryogenic Grinder Inner Bowl
Catalog No. 03391196
Change view
Click to view available options
For Use With (Equipment):
Cryo-Blade
Catalog No. For Use With (Equipment)
03-391-196 Cryo-Blade
1 options
Catalog No. 03-391-196 Supplier Cole-Parmer™ Supplier No. 6121170
Add to Cart
Add to Cart
Optimize performance of your Cole-Parmer™ Cryo-Blade™ Cryogenic Grinder
  • Accessories to rapidly pulverize samples from bulk to analytical preparation
  • Excellent for cryogenic grinding of fruits, vegetables, seeds, stalks, roots, fungi, and flowers
  • Suitable for use with the Cole-Parmer™ CG-900 Cryo-Blade™ cryogenic grinders, 61211-71 and 61211-72. Mills sold separately
Specifications
Product Type Processor Inner Bowl
For Use With (Equipment) Cryo-Blade
Material Metal