Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam cGAMP Disodium Salt, STING Agonist, >98% Purity, 100 μg

Catalog No. 502583268
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3268 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3268 Supplier Abcam Supplier No. AB144865100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent STING agonist that functions as a second messenger to trigger interferon production.

  • Molecular Weight: 674.4 Da
  • Purity: >98%
  • Description: Potent STING agonist that acts as a second messenger, triggering interferon production in response to cytosolic DNA.
  • Activity: Active in vitro; effective concentration in mouse cells at 10 nM.
  • Composition: 100% cGAMP disodium salt.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule STING
Shipping Condition Ambient
Buffer 100% cGAMP disodium salt
Content And Storage Store at -20°C.
Format Vial
Formulation 100% cGAMP disodium salt
Quantity 100 μg
For Use With (Application) STING agonist, Interferon production induction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.