Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam cGAMP Disodium Salt, STING Agonist, >98% Purity, 500 μg
Description
- Molecular weight: 674.4 Da
- Purity: >98%
- Potent STING agonist: Acts as a second messenger triggering interferon production in response to cytosolic DNA.
- Effective concentration: 10 nM in mouse cells.
- Activity: Active in vitro.
- Composition: 100% cGAMP disodium salt.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | STING |
| Buffer | 100% cGAMP disodium salt |
| Content And Storage | Store at -20°C. |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Formulation | 100% cGAMP disodium salt |
| Quantity | 500 μg |
| For Use With (Application) | STING agonist, Interferon production induction |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?