Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGGBP1 Mouse anti-Human, Rat, Clone: 2F5G9, Proteintech
SDP

Catalog No. 501736945 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6945 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6945 Supplier Proteintech Group Inc Supplier No. 665241IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a CGG repeat-binding protein that primarily localizes to the nucleus. CGG trinucleotide repeats are implicated in many disorders as they often act as transcription- and translation-regulatory elements, can produce hairpin structures which cause DNA replication errors, and form regions prone to chromosomal breakage. CGG repeats are also targets for CpG methylation. In addition to its ability to bind CGG repeats and regulate transcription, this gene is believed to play a role in DNA damage repair and telomere protection. In vitro studies indicate this protein does not bind to methylated CpG sequences.

Specifications

Antigen CGGBP1
Applications Western Blot
Classification Monoclonal
Clone 2F5G9
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CGGBP1
Gene Accession No. Q9UFW8
Gene Alias 20 kDa CGG binding protein, CGG binding protein 1, CGGBP, CGGBP1, p20 CGGBP, p20 CGGBP DNA binding protein
Gene Symbols Cggbp1
Host Species Mouse
Immunogen CGGBP1 Fusion Protein Ag1129
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288353, 8545
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.