Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CGGBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592169 Supplier Invitrogen™ Supplier No. PA592169
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MERFVVTAPP ARNRSKTALY VTPLDRVTEF GGELHEDGGK LFCTSCNVVL NHVRKSAISD HLKSKTHTKR KAEFEEQNVR KKQRPLTASL QCNSTAQTEK VSVIQDFVKM CLEANIPLEK ADHPAVRAFL SRHVKNGGSI PKSDQLRRAY LPDGYENENQ LLNSQDC; Positive Samples: Raji, HeLa, 293T, Jurkat; Cellular Location: Nucleus.

CGGBP1 influences expression of the FMR1 gene (MIM 309550), which is associated with the fragile X mental retardation syndrome (MIM 300624), by specifically interacting with the 5-prime (CGG)n-3-prime repeat in its 5-prime UTR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGGBP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Cggbp1
Gene Accession No. Q9UFW8
Gene Alias 20 kDa CGG-binding protein; AA960172; AL023996; AW045841; CGG triplet repeat binding protein 1; CGG triplet repeat-binding protein 1; CGG-binding protein 1; CGGBP; Cggbp1; p20-CGG binding protein; p20-CGGBP; p20-CGGBP DNA-binding protein
Gene Symbols Cggbp1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-167 of human CGGBP1 (NP_0036543).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8545
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.