Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CGI-143 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540483
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540483 Supplier Invitrogen™ Supplier No. PA540483
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AGSGAIGPVE AAIRTKLEEA LSPEVLELRN ESGGHAVPPG SETHFRVAVV Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 93%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 91%.

CGI-143 encodes proteins that seem to be involved in cell proliferation or cell-cycle regulation, but the molecular function is still unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGI-143
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BOLA1
Gene Accession No. Q9Y3E2
Gene Alias 1810037G04Rik; bolA family member 1; bolA homolog 1; Bola1; bolA-like 1; bolA-like 1 (E. coli); bolA-like protein 1; CGI-143; hBolA; hypothetical protein LOC509530; RGD1310586; zgc:101119
Gene Symbols BOLA1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CGI-143 (aa 24-73).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51027
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.