Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CGI 1746
GREENER_CHOICE

Catalog No. p-7223020
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 69-651-0

Supplier: Tocris Bioscience™ 6965/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, reversible BTK inhibitor

CGI 1746 is a potent, reversible inhibitor of Bruton's tyrosine kinase (Btk) (IC50 = 1.9 nM). CGI 1746 decreases cell survival in prostate cancer cell lines. In a cell model, CGI 1746 blocks the production of FcγR-induced cytokines IL-1β, TNFα and IL-6 (IC50 values are 36 nM, 47 nM and 353 nM, respectively).

Specifications

Chemical Name or Material N-[3-[4,5-Dihydro-4-methyl-6-[[4-(4-morpholinylcarbonyl)phenyl]amino]-5-oxo-2-pyrazinyl]-2-methylphenyl]-4-(1,1-dimethylethyl)benzamide
CAS 910232-84-7
Quantity 10 mg
Target Brutons Tyrosine Kinase (BTK) Inhibitors
Molecular Formula C34H37N5O4
Purity 0.97
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.