Learn More
MBL International Corporation CycLex Cyclic GMP dependent protein kinase (cGK) Assay Kit
SDP
Supplier:  MBL International Corporation CY1161
Encompass_Preferred
Size:96 Assays; Target:cGKs; The CycLex Research Product CycLexCyclic GMP dependent protein kinase (cGK) Assay Kit is primarily designed to measure the activities of purified the cGK family of kinases for the rapid and sensitive evaluation of inhibitors or activators. It can be used for 96 assays.CycLex Cyclic GMP dependent protein kinase (cGK) Assay Kit.
Catalog No. 50-455-49
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.