Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva cGMP Direct BioTrak™ EIA

Catalog No. 45001771 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
96 Determinations
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-771 96 Determinations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-771 Supplier Cytiva Supplier No. RPN226
Add to Cart
Add to Cart

Use for direct measurement of cGMP in cell lysates. Cytiva Life Sciences™ cGMP Direct BioTrak™ EIA is a novel lysis reagent that allows cGMP measurement without any need for removal of extraction reagents.

  • Dual range: 2–512 fmol/well [acetylation], 50–12 800 fmol/well [non-acetylation]
  • Sensitivity: 14pg/mL [acetylation], 161pg/mL [non-acetylation]
  • One-day protocol
  • Flexible method-choice of assay protocols
  • Rapid assay protocol
  • Non-radioactive
  • Precise and accurate measurement
  • Ready-to-use substrate
  • Color coded reagents

TRUSTED_SUSTAINABILITY

Specifications

Assay Sensitivity 14pg/mL
Product Type cGMP direct Biotrak EIA
Sufficient For 96-well Microplate
For Use With (Application) Enzyme immunoassay for direct cGMP measurement
Kit Contents Acetylation reagents, Lysis reagents
Quantity 96 Determinations
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.