Learn More
Invitrogen™ cGMP Polyclonal Antibody

Description
PA1-10841 detects cGMP. PA1-10841 has been successfully used in ELISA applications. The PA1-10841 immunogen is Succinylated cGMP, conjugated to BSA.
Specifications
Specifications
| Antigen | cGMP |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 0.1% sodium azide |
| Gene Alias | C10H12N5O7P; Cyclic GMP |
| Host Species | Sheep |
| Immunogen | Succinylated cGMP, conjugated to BSA. |
| Quantity | 200 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.