Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ cGMP Recombinant Rabbit Monoclonal Antibody (RM467)
GREENER_CHOICE

Catalog No. PIMA544557
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544557 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA544557 Supplier Invitrogen™ Supplier No. MA544557
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody reacts to the methoxy group of Polyethylene glycol (PEG). It is specific for methoxypoly(ethylene glycol).

Guanosine cyclic 3',5'-(hydrogen phosphate) is a guanine nucleotide containing one phosphate group which is esterified to the sugar moiety in both the 3'- and 5'-positions. Cyclic gmp is a cellular regulatory agent and has been described as a second messenger. Its levels increase in response to a variety of hormones, including acetylcholine, insulin, and oxytocin and it has been found to activate specific protein kinases. [National Center for Biotechnology Information. PubChem Compound Database; CID=24316].
TRUSTED_SUSTAINABILITY

Specifications

Antigen cGMP
Applications ELISA
Classification Recombinant Monoclonal
Clone RM467
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene Alias C10H12N5O7P; Cyclic GMP
Host Species Rabbit
Immunogen KLH-conjugated cGMP.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.