Learn More
Cayman Chemical CGP 37157, 75450-34-9, 10 mg
SDP
Supplier:  Cayman Chemical 1561110
Encompass_Preferred
Molecular Formula C15H11Cl2NOS, Purity ≥95%, Formula Weight 324.2, Synonyms: 7-chloro-5-(2-chlorophenyl)-1,5-dihydro-4,1-benzothiazepin-2(3H)-one; A selective inhibitor of the mitochondrial sodium-calcium exchanger (IC50 = 0.36 M in isolated mitochondria); commonly used to study the role of mitochondrial-derived calcium in cytoplasmic calcium homeostasis; inhibits mitochondrial calcium efflux via NCLX (IC50 = 5 M).
Catalog No. NC0979196
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.