Learn More
Apexbio Technology LLC CGS 21680 HCl 124431-80-7 50mg
SDP
Supplier:  Apexbio Technology LLC B132450
Encompass_Preferred
CGS 21680 HCl is a selective agonist of the adenosine A2 receptor subtype which exerts its pharmacological effects through specific binding and activation of A2 receptors It demonstrates potent affinity for A2 receptors evidenced by its reported IC50 of approximately 22 nM in radioligand binding assays performed on rat striatal membranes It shows minimal interaction with other neurotransmitter binding sites such as adrenergic dopaminergic or serotonergic receptors In animal models administration of CGS 21680 HCl induces coronary vasodilation reduces blood pressure and exhibits transient increases in heart rate Therefore CGS 21680 HCl is frequently employed in cardiovascular and neurological research to evaluate adenosine receptor-mediated physiological processes and receptor signaling pathways
Catalog No. 50-000-62117
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.