Learn More
Apexbio Technology LLC CH5132799, 5mg. Cas: 1007207-67-1 MFCD: MFCD22419020. Class I PI3K inhibitor
SDP
Supplier:  Apexbio Technology LLC B21835
Encompass_Preferred
CH5132799 is an inhibitor of class I phosphatidylinositol 3-kinase (PI3K) with IC50 value of 14nM against PI3K [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5327
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.