Learn More
Medchemexpress LLC HY-135299 5mg Medchemexpress, CH6953755 CAS:2055918-71-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1352995MG
Encompass_Preferred
Medchemexpress, HY-135299 5mg CH6953755 CAS:2055918-71-1 CH6953755 is a potent, orally active and selective YES1 kinase (a member of the SRC family) inhibitor with an IC50 of 1.8 nM. CH6953755 inhibits YES1 kinase, leading to antitumor activity against YES1 Gene -amplified cancers in vitro and in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0332
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.