Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHADL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572888
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572888 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572888 Supplier Invitrogen™ Supplier No. PA572888
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHADL Polyclonal Antibody for Western Blot, IHC, ELISA

Potential negative modulator of chondrocyte differentiation. Inhibits collagen fibrillogenesis in vitro. May influence chondrocyte's differentiation by acting on its cellular collagenous microenvironment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHADL
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CHADL
Gene Accession No. E9Q7T7, Q6NUI6
Gene Alias AY100452; Chadl; chondroadherin like; chondroadherin pseudogene; chondroadherin-like; chondroadherin-like protein; chondroadherin-like protein-like; D930017K21Rik; leucine rich repeat containing 15 pseudogene; SLRR4B
Gene Symbols CHADL
Host Species Rabbit
Immunogen CHADL antibody for a 19 amino acid peptide from near the carboxy terminus of human CHADL. The immunogen is within the last 50 amino acids of CHADL.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100910434, 150356, 214685
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.