Learn More
Eisco™ Chain Drive with Tensioner Mechanical Demonstration Model
SCI_ED
Catalog No. S27670 Shop All Eisco Products
Change view
Click to view available options
Chain Drive with Tensioner Mechanical Demonstration Model
Catalog No.
S27670 Chain Drive with Tensioner Mechanical Demonstration Model
1 options
Catalog No. S27670 Supplier Eisco™ Supplier No. PH0366TM36
The Chain Drive with Tensioner is an interactive visual demonstration of a mechanical power transmission system that uses a chain to transfer torque between rotating shafts and a component to maintain proper tension in the chain.
  • Hand-cranked interactive model for visual learning.
  • Shows the fundamentals of how a chain drive with tensioner works.
  • Open system is sandwiched by two clear plastic plates for unobstructed study.
  • Constructed with heavy duty metal and plastic.
  • Includes comprehensive instruction and experiment manual.
Specifications
Product Type Chain Drive with Tensioner Mechanical Demonstration Model