Learn More
Ampco Chain Pipe Wrench
SDP
Catalog No. 19174699 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Tubes of up to 12 in. Dia
Tubes of up to 4 in. Dia
Tubes up to 1 and 1/2 in. Dia
Tubes up to 2 and 1/2 in. Dia
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-174-699 Tubes up to 2 and 1/2 in. Dia
19-174-695 Tubes up to 1 and 1/2 in. Dia
19-174-701 Tubes of up to 12 in. Dia
19-174-703 Tubes of up to 4 in. Dia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-174-699 Supplier Ampco Supplier No. W61
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant wrench. Made in the USA.
  • Robust, adjustable tools designed specifically to grip and turn large-diameter pipes or other cylinders.
  • With high torque, these chain pipes are designed for tough jobs.
Specifications
Product Type Chain Pipe Wrench
For Use With (Equipment) Tubes up to 2 and 1/2 in. Dia
Length (English) 27 in.
No. per Pack 1 Ea.