Learn More
Market Lab Chameleon Cooler, 4""""L x 4""""W x 4""""H
Supplier:  Market Lab 8320
Encompass_Preferred
Maintain a temperature of 32°–34°F for over six hours with Chameleon Cooler, keeping up to six tubes chilled and preventing the metabolism of cells in blood. Simply place the cooler in the freezer overnight before use.
Catalog No. NC9811696
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.