Learn More
FUJIFILM IRVINE SCIENTIFIC INC CHANG MARROW
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 91031100ML
Encompass
CHANG Marrow is a complete, ready-to-use medium optimized to support bone marrow cultures for karyotyping and other genetic testing of various hematological disorders. After an extensive optimization process to create a superior, innovative blend of growth factors, Irvine Scientific's CHANG Marrow has been shown to out-perform other commercially available media in clinical studies. Packaged in single bottles, ready-to-use with no mixing required. For in vitro diagnostic use. 100 mL.
Catalog No. NC0985123
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.