Learn More
Honeywell™ Chargers, Batteries, and Power Accessories for PA700 PAPR
Choose among a variety of chargers, batteries, and accessories designed to complement the PA700 PAPR system.
Supplier: Honeywell™ PA720
This item is not returnable.
View return policy
Description
- Replacement components for the PA700 Powered Air-Purifying Respirator
- Rechargeable battery pack can power a PAPR for an entire 8-hour work shift
- Smart charger system can re-charge a fully depleted battery in approximately 4.5 hours
- Choose from 1 bay or 4 bay charger dock
- Battery Pack Features an on-board, state-of-charge display, which allows the user to evaluate the amount of remaining charge in the pack, without installing it into the blower
- Charger kit continuously monitors the health of the battery pack and alerts the user to a potential problem via an LED display panel
Specifications
Lithium-ion | |
Reusable | |
Primair™ 700 Series | |
PA720 |