Learn More
Scott Safety™ Charging Systems for Pak-Tracker Firefighting Locator System
Catalog No. 19824455
Change view
Click to view available options
Product Type:
Desktop Charging System
Truck Charging System
Voltage:
12 V
110 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Voltage
19-824-455 Truck Charging System 12 V
19-824-456 Desktop Charging System 110 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-824-455 Supplier Scott Safety™ Supplier No. 7012471473
Add to Cart
Add to Cart
Truck charging system chargers or recharging the nickel-metal hydride battery pack
Specifications
Product Type Truck Charging System
Voltage 12 V