Learn More
Cayman Chemical chartreusIs A23187 1mg
SDP
Supplier:  Cayman Chemical 110161 MG
Encompass_Preferred
Immunogen Peptide from the C-terminal region of rat RSK2 Host rabbit Species Reactivity ( ) rat RSK2 Application(s) WB RSKs 1-4 are downstream members of the ERK/MAPK cascade Recent work suggests that RSK2 exerts a tonic regulation on G protein-coupled signaling
Catalog No. 50-295-2681
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.