Learn More
Enzo Life Sciences Charybdotoxin (recombinant) (100µg). CAS: 95751-30-7
Supplier: Enzo Life Sciences BMLKC1680100
Charybdotoxin was originally isolated from the venom of the Israeli scorpion Leiurus quinquestriatus hebraeus. Charybdotoxin blocks KCa1.1 (large conductance Ca2+ activated K+, Slo) channels in nanomolar concentrations as as well as Kv1.2 (Kd, 14 nM) and Kv1.3 (Kd, 2.6 nM) channels.). However, experiments with cloned KCa1.1 channels demonstrate the strong effect of the slob subunits on the potency of block by Charybdotoxin. This product is a recombinant peptide expressed in and extracted from E. coli. Purity: ≥98%. Solubility: Soluble in Water. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.