Learn More
Description
- Wide mediumweight roll for absorbing everyday spills, leaks and overspray around machines or in large areas
- Bright yellow color stands out for workers to see hazards, repairs or cleanups in progress
- Durable top layer resists scuffs and stands up to foot traffic without creating a trip hazard
- Eye-catching safety message repeats throughout the roll
- Easy-tear perforations let you tear off the size needed
- Absorbs most common industrial liquids, including oils, coolants, solvents, water, etc.
- Can be wrung out and incinerated after use to reduce waste or for fuels blending
- 24 in. x 75 ft. roll
Specifications
Specifications
| Absorbancy | 13.5 gal. |
| Design | Printed with 'Don't Let A Drip Turn Into A Slip' |
| Material | Polypropylene |
| Type | Absorbent Roll |
| Disposable | Single-use |
| Weight Capacity | Medium-weight |
| Length (English) | 75 ft. |
| Length (Metric) | 22.9 m |
| Width (English) | 24 in. |
| Width (Metric) | 61 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
