Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIG™ Chat Mat™ High-Visibility Absorbent Roll
SDP

Catalog No. 18366603 Shop All New Pig Corporation Products
Change view
Click to view available options
Design:
Printed with 'Don't Let A Drip Turn Into A Slip'
Printed with 'Mom Doesn't Work Here/Clean Up'
Printed with 'Safety Is Everyone's Business'
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Design
18-366-603 Printed with 'Safety Is Everyone's Business'
18-366-601 Printed with 'Don't Let A Drip Turn Into A Slip'
18-366-602 Printed with 'Mom Doesn't Work Here/Clean Up'
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-366-603 Supplier PIG™ Supplier No. MAT60781
Add to Cart
Add to Cart

High-visibility yellow mat roll that features a repeating safety sign image.

  • Wide mediumweight roll for absorbing everyday spills, leaks and overspray around machines or in large areas
  • Bright yellow color stands out for workers to see hazards, repairs or cleanups in progress
  • Durable top layer resists scuffs and stands up to foot traffic without creating a trip hazard
  • Eye-catching safety message repeats throughout the roll
  • Easy-tear perforations let you tear off the size needed
  • Absorbs most common industrial liquids, including oils, coolants, solvents, water, etc.
  • Can be wrung out and incinerated after use to reduce waste or for fuels blending
  • 24 in. x 75 ft. roll

Specifications

Absorbancy 13.5 gal.
Design Printed with 'Safety Is Everyone's Business'
Material Polypropylene
Type Absorbent Roll
Disposable Single-use
Weight Capacity Medium-weight
Length (English) 75 ft.
Length (Metric) 22.9 m
Width (English) 24 in.
Width (Metric) 61 cm
Weight (English) 11 lb.
Weight (Metric) 5 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.