Learn More
Description
Immunogen sequence: GLFSSYRLPG HTQDTLVAQN SSIMPEPEHV IVACCNQFFV LDVVINFRRL SEGDLFTQLR KIVKMASNED ERLPPIGLLT SDGRSEWAEA RTVLVKDSTN Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.
Specifications
Specifications
| Antigen | ChAT |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL3169 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CHAT |
| Gene Accession No. | P28329, P32738, Q03059 |
| Gene Alias | acetyl CoA; acetyl CoA:choline O-acetyltransferase; B230380D24Rik; CHAT; CHOACTASE; choline acetylase; choline acetyltransferase; choline acetyltransferase (EC 2.3.1.6); choline acetyltransferase long variant; choline acetyltransferase short variant; choline O-acetyltransferase; CMS1A; CMS1A2; CMS6; LOW QUALITY PROTEIN: choline O-acetyltransferase; N-type choline acetyltransferase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
