Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHD1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555940
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555940 Supplier Invitrogen™ Supplier No. PA555940
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSRDQEEGKN HMYLFEGKDY SKEPSKEDRK SFEQLVNLQK TLLEKASQEG RSLRNKGSVL IPGLVEGSTK RKRVLSPEEL EDRQKKRQEA AAKRRRLIEE KKR Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 78%.

In response to DNA strand breaks, chromatin adopts a relaxed structure due to the addition of poly(ADP-ribose) (PAR) to chromatin proteins by PARP enzymes (see PARP1; MIM 173870), and this relaxation facilitates the repair of DNA damage. CHD1L interacts with PAR and has a role in chromatin relaxation following DNA damage (Ahel et al., 2009 [PubMed 19661379]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD1L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHD1L
Gene Accession No. Q86WJ1
Gene Alias 4432404A22Rik; ALC1; amplified in liver cancer 1; amplified in liver cancer protein 1; CHD1L; CHDL; chromodomain helicase DNA binding protein 1 like; chromodomain helicase DNA binding protein 1-like; chromodomain-helicase-DNA-binding protein 1-like; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 1-like; Snf2p; Unknown (protein for MGC:128792)
Gene Symbols CHD1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHD1L. Recombinant protein control fragment (Product #RP-94193).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9557
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.