Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD3 Antibody, ProSci
SDP

Catalog No. 16815371 Shop All ProSci Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-815-371 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-815-371 Supplier ProSci LLC Supplier No. 185820100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

CHD3 Polyclonal Antibody specifically detects COX5B in Human, Mouse, Rat Samples. It is validated for Western Blot.

Specifications

Antigen CHD3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution WB: 1:200 - 1:1000
Formulation PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
Gene Alias Mi-2a, Mi2-ALPHA, SNIBCPS, ZFH, chromodomain-helicase-DNA-binding protein 3, ATP-dependent helicase CHD3, CHD-3, hZFH, mi-2 autoantigen 240 kDa protein, zinc finger helicase, zinc-finger helicase (Snf2-like)
Host Species Rabbit
Purification Method Affinity purification
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1107
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze / thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Research Category Immunology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.