Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD5 Antibody, Biotin, R&D Systems™
SDP

Catalog No. NB134645
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB134645 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB134645 Supplier R&D Systems Supplier No. NBP306267B

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

CHD5 Polyclonal specifically detects CHD5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence/Immunohistochemistry/Immunohistochemistry (Paraffin).

Specifications

Antigen CHD5
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ATP-dependent helicase CHD5, CHD-5, chromodomain helicase DNA binding protein 5, chromodomain-helicase-DNA-binding protein 5, DKFZp434N231, EC 3.6.1, EC 3.6.4.12, KIAA0444
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CHD5 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26038
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.