Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD9/CReMM Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501559829
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9829 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9829 Supplier Bethyl Laboratories, Inc Supplier No. A301226AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

Acts as a transcriptional coactivator for PPARA and possibly other nuclear receptors. Proposed to be a ATP-dependent chromatin remodeling protein. Has DNA-dependent ATPase activity and binds to A/T-rich DNA. Associates with A/T-rich regulatory regions in promoters of genes that participate in the differentiation of progenitors during osteogenesis (By similarity).

Specifications

Antigen CHD9/CReMM
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CHD9
Gene Accession No. Q3L8U1
Gene Alias AD013, ATP-dependent helicase CHD9, BC022889, CHD-9, chromatin remodeling factor CHROM1, chromatin-related mesenchymal modulator, chromatin-remodeling factor CHROM1, ciprofibrate bound protein p240, CReMM, FLJ12178, KIAA0308, KISH2, kismet homolog 2, peroxisomal proliferator-activated receptor A-interacting complex 320 kDa protein, PPAR{gamma}-interacting cofactor 320 kDa, PPAR-alpha-interacting complex protein 320 kDa, PRIC320, proteinx0008
Gene Symbols CHD9
Host Species Rabbit
Immunogen Between 25 and 75
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80205
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.