Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig Checkered Carpet Adhesive Backed Grippy Floor Mat - Gray
SDP

Catalog No. 18813371 Shop All New Pig Corporation Products
Change view
Click to view available options
Dimensions (L x W):
73 x 6.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18-813-371 73 x 6.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-813-371 Supplier New Pig Supplier No. GRP926X10GY

Indoor use on concrete, vinyl, linoleum, tile, and laminate flooring, 6 ft. x 10 ft. (W x L), 1 roll.

  • Adhesive-backed floor mat
  • Dries out quickly without moisture soaking through to the floor; absorbs water, oil and grease and traps dirt and salt to keep it from being tracked around
  • Stays in place for up to 3 to 6 months, depending on conditions and traffic patterns
  • Easy to clean: you can sweep, vacuum, mop, use your shop/wet vac or run your floor scrubber over Grippy Mat and it won't budge
  • Thin, no rippled edges to obstruct doors or create tripping hazards.
  • ADA compliant for wheelchairs and walkers; carts and pallet jacks roll right over it
  • Sticks to most commercial floor surfaces including concrete, vinyl, linoleum, ceramic tile, quarry tile and laminate flooring

Specifications

Dimensions (L x W) 73 x 6.5 in.
Length (Metric) 185 cm
Width (English) 6.5 in.
Length (English) 73 in.
Material Polyester
Width (Metric) 17 cm
Color Smoke Gray
Type Adhesive Floor Mat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.