Learn More
New Pig Checkered Carpet Adhesive Backed Grippy Floor Mat - Gray
Shop All New Pig Corporation Products

Description
- Adhesive-backed floor mat
- Dries out quickly without moisture soaking through to the floor; absorbs water, oil and grease and traps dirt and salt to keep it from being tracked around
- Stays in place for up to 3 to 6 months, depending on conditions and traffic patterns
- Easy to clean: you can sweep, vacuum, mop, use your shop/wet vac or run your floor scrubber over Grippy Mat and it won't budge
- Thin, no rippled edges to obstruct doors or create tripping hazards.
- ADA compliant for wheelchairs and walkers; carts and pallet jacks roll right over it
- Sticks to most commercial floor surfaces including concrete, vinyl, linoleum, ceramic tile, quarry tile and laminate flooring
Specifications
Specifications
| Dimensions (L x W) | 36 x 8.5 in. |
| Length (English) | 36 in. |
| Width (English) | 8.5 in. |
| Color | Smoke Gray |
| Length (Metric) | 91 cm |
| Material | Polyester |
| Type | Adhesive Floor Mat |
| Width (Metric) | 22 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.