Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine-Free/Total
Catalog No. 09053103 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0 to 4mg/L
0.02 to 0.3mg/L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
09-053-103 0.02 to 0.3mg/L
09-053-106 0 to 4mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-053-103 Supplier Lovibond™ Supplier No. 147000
Add to Cart
Add to Cart
Includes
  • Aqua Comparator Housing (D55 for L147000)
  • Aqua Comparator Disc
  • DPD No. 1 Rapid Tablets
  • DPD No. 3 Rapid Tablets
  • 2 x 13.5mm Sample Cells
  • Tablet Crusher
  • Instructions
  • Carrying case
Accurate test kits for routine water analysis
  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
Accuracy ±5%
For Use With (Application) Routine water analysis
Includes Aqua comparator™ housing, aqua comparator™ disc D55, DPD No. 1 tablets, DPD No. 3 tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
No. of Tests 100
Range 0.02 to 0.3mg/L
Type Chlorine Free and Total Test Kit