Learn More
Cayman Chemical Cheldonn 25mg
SDP
Supplier:  Cayman Chemical 2509725 MG
Encompass_Preferred
An NSAID and COX inhibitor (IC50s 3 and 9.3 UM for human COX-1 and COX-2 respectively) inhibits TNF-a-induced increases in COX-2 levels and NF-KB activation in HT-29 colon cancer cells in a concentration-dependent manner inhibits fMLP- or A23187-induced calcium influx in PMN (IC50s 29 and 14 UM respectively) activates various ion channels including TRPC6 and KCa1.1 inhibits various ion channels including TRPC3 and CFTR reduces increases in intestinal fluid secretion and intestinal barrier disruption in mice infected with the El Tor variant of V. cholerae at 20 mg/kg
Catalog No. 50-293-4470
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.