Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chelidamic Acid Monohydrate 95.0+%, TCI America™
SDP

Catalog No. C082125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C082125G 25 g
C08215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C082125G Supplier TCI America Supplier No. C082125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 138-60-3
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
MDL Number MFCD00150943
InChI Key XTLJJHGQACAZMS-UHFFFAOYSA-N
Synonym chelidamic acid, 4-hydroxypyridine-2,6-dicarboxylic acid, 4-oxo-1,4-dihydropyridine-2,6-dicarboxylic acid, 4-hydroxy-2,6-pyridinedicarboxylic acid, chelidamic acid hydrate, 2,6-pyridinedicarboxylic acid, 1,4-dihydro-4-oxo, 2,6-dicarboxy-4-hydroxypyridine, 1,4-dihydro-4-oxopyridine-2,6-dicarboxylic acid, 2,6-pyridinedicarboxylic acid, 4-hydroxy, 4-oxo-1,4-dihydro-2,6-pyridinedicarboxylic acid
PubChem CID 8743
IUPAC Name 4-oxo-1,4-dihydropyridine-2,6-dicarboxylic acid
SMILES OC(=O)C1=CC(=O)C=C(N1)C(O)=O

Specifications

Melting Point 267°C
Color White-Yellow
Quantity 25 g
Formula Weight 183.12
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chelidamic Acid Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.