Learn More
Cole-Parmer ICI Quick Ship CHEM Exhaust Snorkel, 57.1" (145 cm) Radius
Supplier:  Cole-Parmer 7898117
Encompass_Preferred
These lightweight, flexible exhaust snorkels with three adjustable, composite friction joints allow you to get close to the source of fumes and particles safely. The two outermost arm joints can rotate and swivel 360 degrees. Constructed of anodized aluminum, the snorkels offer strength, durability, and ease of handling. The arms are 3” in diameter and are available with a maximum working radius of 42.9” (109 cm) or 57.1” (145 cm). Each arm comes with a 5.5” mini hood that will accept optional hoods.
Catalog No. NC2103207
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.