Learn More
Chemglass Life Sciences Temperature Monitor Only, Digital, PT-100, 120V
SDP
Supplier:  Chemglass Life Sciences CG3498P100
Encompass_Preferred
Temperature monitor features a built in audible alarm, PT-100 probe receptacle, and a bright LED display for easy viewing. The monitor is contained in a compact stainless steel cabinet with a side-mounted clamp for securing to a 1/2" diameter support rod. It has a separate ON/OFF switch and a circuit breaker. Dimensions: 2. 6" High x 3. 2" Wide x 5. 5" Deep. PLEASE NOTE: SUPPORT ROD, CG-3499-A-20 PROBE ADAPTER CABLE AND CG-1978-P PROBE ARE SOLD SEPARATELY.
Catalog No. 50-273-3530
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.