Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Chemerin-9, Human
GREENER_CHOICE

Catalog No. 71161
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-161 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 71-161 Supplier Tocris Bioscience™ Supplier No. 7116/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent CMKLR1 agonist; c-terminal peptide of full length human chemerin

Chemerin-9, Human is a potent chemokine-like receptor 1 (CMKLR1; also known as chemerin receptor 23, ChemR23) agonist (EC50 = 7.1 nM at CMKLR expressed in CHO cells). Corresponds to C-terminal of full length human Chemerin, amino acids 149 - 157. Activates Gi/o signaling pathways in vitro; inhibits cAMP production and promotes phospholipase C production and Ca2+ mobilization.

Specifications

Purity 95%
Content And Storage Store at -20°C
Molecular Weight (g/mol) Observed: 1063.18
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.