Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Chemical, Biohazard & Hazardous Material Label - Self-Sticking Polyester

Catalog No. 19106759 Shop All Brady Corporation Products
Change view
Click to view available options
Display Text:
ACETYLENE
CHAIN CYLINDERS
CORROSIVE MATERIALS
DANGER DIESEL FUEL
DIESEL FUEL
FLAMMABLE GAS
FLAMMABLE MATERIALS
GASOLINE
LEAD WORK AREA POISON NO SMOKING OR EATING
OXYGEN
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
19-106-759 GASOLINE
19-106-760 OXYGEN
19-106-758 DIESEL FUEL
19-107-580 FLAMMABLE MATERIALS
19-107-581 DANGER DIESEL FUEL
19-107-582 FLAMMABLE GAS
19-107-584 LEAD WORK AREA POISON NO SMOKING OR EATING
19-107-583 CORROSIVE MATERIALS
19-106-757 ACETYLENE
19-106-782 CHAIN CYLINDERS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 19-106-759 Supplier Brady™ Supplier No. 16876
Add to Cart
Add to Cart

Self-Sticking Polyester: Self-Sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.

  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704; OSHA 1910.1200
Description Label
Adhesive Type Permanent Acrylic, Self-adhesive
Color Red on White
Finish Gloss
Format Label
Material Polyester
Printable Sides Single-side Printable
Product Type Label
Display Text GASOLINE
Languages English
Quantity 1 Ea.
Shape Rectangle
Size (English) 5 x 3.5 in. (W x H)
Dimensions (L x W) 3.5 x 5 in.
Width (English) 5 in.
Width (Metric) 127 mm
Height (English) 3.5 in.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.