Learn More
Brady™ Chemical, Biohazard & Hazardous Material Sign - Rigid Polyethylene
Catalog No. 19106777 Shop All Brady Corporation Products
Catalog No. 19-106-777 Supplier Brady™ Supplier No. 17586
Rigid Polyethylene: Rigid polyethylene (B-450) is ideal for high visibility fire equipment signs and a variety of custom markers and signs.
  • 3 to 5 year average outdoor durability
  • Linear polyethylene sheet contains UV stabilizers
  • Surface printing holds up to most common chemicals, oils and water
  • Service temperature of -40°F to 180°F (-40°C to 82°C)
Specifications
Product Type Chemical, Biohazard, and Hazardous Material Sign
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.